missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alisol F, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Alisol F is a triterpene isolated from Alisma orientalis, has immunosuppressive and anti-virus functions. Alisol F exhibits inhibitory activity in vitro on hepatitis B virus (HBV) surface antigen (HBsAg) secretion of the HepG2.2.15 cell line with an IC50 of 0.6 μM.
Specifications
Specifications
| Chemical Name or Material | Alisol F |
| Physical Form | Solid |
| CAS | 155521-45-2 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C30H48O5 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : 100 mg/mL (204.62 mM; Need ultrasonic) |
| SMILES | C[C@]1(C(C[C@@H]2O)=C([C@@H]3C)[C@](O[C@@]([C@@H](O)C(C)(O)C)([H])C3)([H])C1)[C@]([C@]2([H])[C@]4(CCC5=O)C)(CC[C@@]4([H])C5(C)C)C |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction