Learn More
Alisol B 23-acetate, MedChemExpress
Product Code. 16489108
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
16489108 10 mg 10mg
16444897 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16489108 Leverantör Medchem Express Leverantörsnummer HYN0805
Please to purchase this item. Need a web account? Register with us today!
Denna artikel kan inte returneras. Se returpolicy
MedChemExpress Alisol B 23-acetate (23-Acetylalismol B), a natural triterpenoid, produces protective effects against EE-induced cholestasis, due to FXR-mediated gene regulation.
Alisol B 23-acetate (23-Acetylalismol B), a natural triterpenoid, produces protective effects against EE-induced cholestasis, due to FXR-mediated gene regulation.
Specifikationer
Chemical Name or Material Alisol B 23-acetate
CAS 26575-95-1
Color White
Purity Grade Notes Research
Molecular Formula C32H50O5
Quantity 10 mg
Synonym 23-Acetylalismol B 23-O-Acetylalisol B Alisol B monoacetate
Solubility Information DMSO : 50 mg/mL (97.14 mM; Need ultrasonic)
SMILES C[C@]([C@@]1(C2=C([C@H](C)C[C@@H]([C@]3([H])C(C)(C)O3)OC(C)=O)CC1)C)(CC[C@@]4([H])C5(C)C)[C@]([C@H](C2)O)([H])[C@]4(CCC5=O)C
Molecular Weight (g/mol) 514.74
Formula Weight 514.74
Percent Purity 98.33%
Grade Research
Physical Form Solid
Visa mer Visa mindre
Research purposes only
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.