Learn More
Aliskiren hemifumarate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Aliskiren (CGP 60536
Specifications
Specifications
| Chemical Name or Material | Aliskiren hemifumarate |
| CAS | 173334-58-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C64H110N6O16 |
| Quantity | 50 mg |
| Synonym | CGP 60536 hemifumarateCGP60536B hemifumarateSPP 100 hemifumarate |
| Solubility Information | H2O : ≥ 50 mg/mL (41.00 mM) |
| SMILES | O=C(O)/C=C/C(O)=O.O=C(NCC(C)(C(N)=O)C)[C@@H](C[C@@H]([C@H](C[C@@H](C(C)C)CC1=CC=C(OC)C(OCCCOC)=C1)N)O)C(C)C.O=C(NCC(C)(C(N)=O)C)[C@H](C(C)C)C[C@H](O)[C@@H](N)C[C@@H](C(C)C)CC2=CC=C(OC)C(OCCCOC)=C2 |
| Molecular Weight (g/mol) | 1219.59 |
| Show More |