missing translation for 'onlineSavingsMsg'
Learn More

Alectinib Hydrochloride, MedChemExpress

Product Code. 16475478
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475478 10 mg 10mg
16491056 200 mg 200mg
16466066 100 mg 100mg
16441657 50 mg 50mg
16428197 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16475478 Supplier Medchem Express Supplier No. HY13011A
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Alectinib Hydrochloride (CH5424802 Hydrochloride; RO5424802 Hydrochloride; AF-802 Hydrochloride) is a potent, selective, and orally available ALK inhibitor with an IC50 of 1.9 nM and a Kd value of 2.4 nM (in an ATP-competitive manner), and also inhibits ALK F1174L and ALK R1275Q with IC50s of 1 nM and 3.5 nM, respectively. Alectinib demonstrates effective central nervous system (CNS) penetration.

Alectinib Hydrochloride (CH5424802 Hydrochloride

Specifications

Chemical Name or Material Alectinib Hydrochloride
CAS 1256589-74-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H35ClN4O2
Quantity 10 mg
Synonym CH5424802 HydrochlorideRO5424802 HydrochlorideAF-802 Hydrochloride
Solubility Information DMSO : 5 mg/mL (9.63 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES [H]Cl.N#CC1=CC2=C(C3=C(N2)C(C)(C4=CC(N5CCC(CC5)N6CCOCC6)=C(C=C4C3=O)CC)C)C=C1
Molecular Weight (g/mol) 519.08
Formula Weight 519.08
Percent Purity 98.06%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.