Learn More
Aldolase, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| CAS | 9024-52-6 |
| Color | White |
| Quantity | 100 mg |
| Synonym | D -Fructose-1, 6-bisphosphate-D -glyceraldehyde-3-phosphate lyase, E.C. 4.1.2.13 |
| Formula Weight | ≈161,000 |
| Physical Form | Powder |
| Chemical Name or Material | Aldolase |