missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDOB Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00 € - 349.00 €
Specifications
| Antigen | ALDOB |
|---|---|
| Clone | 3D2E4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234799
|
Proteintech
68396-1-IG-20UL |
20 μL |
145.00 € 129.00 € / 20µL Save 16.00 € 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237776
|
Proteintech
68396-1-IG-150UL |
150 μL |
410.00 € 349.00 € / 150µL Save 61.00 € 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ALDOB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ALDOB. This antibody recognizes human, mouse, rat, rabbit antigen. The ALDOB antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| ALDOB | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 229 | |
| IgG2a | |
| Protein A purification | |
| Monoclonal Antibody |
| 3D2E4 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Rabbit | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title