Learn More
Aldehyde Oxidase 1/AOX1 Goat anti-Human, Polyclonal, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF5928-SP
Encompass
Product Code. 15758902
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
Aldehyde Oxidase 1/AOX1 Polyclonal specifically detects Aldehyde Oxidase 1/AOX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.