Learn More
Aldehyde Oxidase 1/AOX1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5928-SP
This item is not returnable.
View return policy
Description
Aldehyde Oxidase 1/AOX1 Polyclonal specifically detects Aldehyde Oxidase 1/AOX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Product Title