missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ALCAM/CD166 Monoclonal antibody specifically detects ALCAM/CD166 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
Specifications
| Antigen | ALCAM/CD166 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 21B9 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 |
| Formulation | PBS, pH 7.4, 150mM NaCl and 50% glycerol |
| Gene Alias | activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from Human ALCAM/CD166 [UniProt Q13740] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?