Learn More
Albendazole-d3 (Methoxy-d3), TRC
Specifications
Specifications
| Chemical Name or Material | Albendazole methoxy-d3 |
| CAS | 1353867-92-1 |
| Molecular Formula | C12 H15 N3 O2 S |
| Quantity | 10 mg |
| Synonym | Trideuteriomethyl N-(5-propylsulfanyl-1H-benzimidazol-2-yl)carbamate |
| InChI Formula | InChI=1S/C12H15N3O2S/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)/i2D3 |
| SMILES | [2H]C([2H])([2H])OC(=O)Nc1nc2cc(SCCC)ccc2[nH]1 |
| IUPAC Name | trideuteriomethyl N-(5-propylsulfanyl-1H-benzimidazol-2-yl)carbamate |
| Molecular Weight (g/mol) | 268.35 |
| Formula Weight | 268.1073 |