missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alanyl tRNA synthetase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31706
This item is not returnable.
View return policy
Description
Alanyl tRNA synthetase Polyclonal specifically detects Alanyl tRNA synthetase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Alanyl tRNA synthetase | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| alanine tRNA ligase 1, cytoplasmic, Alanine--tRNA ligase, alanyl-tRNA synthetase, alanyl-tRNA synthetase, cytoplasmic, AlaRS, CMT2N, EC 6.1.1, EC 6.1.1.7, Renal carcinoma antigen NY-REN-42 | |
| Rabbit | |
| 107 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.23mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P49588 | |
| AARS1 | |
| Recombinant protein encompassing a sequence within the center region of human alanyl-tRNA synthetase. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 16 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction