Learn More
Alanine-d4, TRC
Product Code. 30318406
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318406 1 g 1g
30313764 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318406 Supplier Toronto Research Chemicals Supplier No. TRCA4814021G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Alanine-d4
CAS 53795-92-9
Molecular Formula C3 D4 H3 N O2
Quantity 1 g
Synonym DL-Alanine-2,3,3,3-d4, DL-Alanine-d4, (R,S)-Alanine-d4, (±)-2-Aminopropionic Acid-d4, (±)-Alanine-d4, DL-Ala-d4, DL-α-Alanine-d4, DL-α-Aminopropionic Acid-d4, NSC 7602-d4, dl-2-Aminopropanoic Acid-d4, dl-Alanine-d4
InChI Formula InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/i1D3,2D
SMILES [2H]C([2H])([2H])C([2H])(N)C(=O)O
IUPAC Name 2-amino-2,3,3,3-tetradeuteriopropanoic acid
Molecular Weight (g/mol) 93.12
Formula Weight 93.0728