Learn More
Al-23 Tube, Both ends open (Thin walls), O.D.: 20mm, I.D.: 19mm, Thermo Scientific™
Product Code. 11358267
Click to view available options
Quantity:
100 mm
200 mm
300 mm
Unit Size:
100mm
200mm
300mm
Product Code. 11358267
Brand: Thermo Scientific Alfa Aesar 031712.DM
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open (Thin walls)
Quantity 100 mm