Learn More
Al-23 Tube, Both Ends Open (Thin Wall);OD (mm), 8;ID (mm), 7
Product Code. p-4905288
Click to view available options
Quantity:
100 mm
200 mm
300 mm
Unit Size:
100mm
200mm
300mm
Product Code. 11388247
Brand: Thermo Scientific Alfa Aesar 031708.DM
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23
Name Note Both ends open (Thin walls)
Quantity 100 mm
Form Tube
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.