Learn More
Al-23 Tube (both ends open);OD (mm), 4;ID (mm), 2
Product Code. 11373917
Change view
Click to view available options
Quantity:
100 mm
200 mm
300 mm
600 mm
800 mm
1000 mm
Unit Size:
1000mm
100mm
200mm
300mm
600mm
800mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373917 300 mm 300mm
11383917 100 mm 100mm
11343917 200 mm 200mm
11353917 600 mm 600mm
11363917 800 mm 800mm
11333917 1000 mm 1000mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11373917 Supplier Thermo Scientific Alfa Aesar Supplier No. 031554.DW
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open
Quantity 300 mm
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.