missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Akti-1/2

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Akti-1/2 is a potent and selective dual Akt1 and 2 inhibitor (IC50 values are 50 and 210 nM, respectively). Selective for Akt1 and 2 over a panel of other tyrosine and serine/threonine kinases. Sensitizes LnCaP cells to TRAIL (TNF-related apoptosis-inducing ligand) induced apoptosis. Also enhances CAR and TCR retroviral transduction of human T cells. Active in vivo.
Specifications
Specifications
| Chemical Name or Material | 1,3-Dihydro-1-[1-[[4-(6-phenyl-1 H-imidazo[4,5-g]quinoxalin-7-yl)phenyl]methyl]-4-piperidinyl]-2 H-benzimidazol-2-one |
| CAS | 612847-09-3 |
| Synonym | Akt Inhibitor VIII |
| Quantity | 10 mg |
| Target | Akt (Protein Kinase B) Inhibitors |
| Molecular Formula | C34H29N7O |
| Formula Weight | 551.64 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction