Learn More
Eurofins DiscoverX Akt3/PKBγ (S472D) Protein, Active
Description
AKT (also called protein kinase B) is a serine/threonine protein kinase that has been implicated in mediating a variety of biological responses including inhibiting apoptosis and stimulating cellular growth. Three mammalian isoforms (AKT1, AKT2, and AKT3) of the kinase have been identified.
EMBL AF124141. The recombinant protein contains an amino acid substitution, S472D, with reference toEMBL AF124141. This mutation mimics phosphorylation of S472.
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl, pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 43 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥75% |
| Protein | PKB gamma (117-end, S472D), active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.