Learn More
STEMCELL Technologies AKT Inhibitor VIII
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- AKT Inhibitor VIII is a cell-permeable, allosteric inhibitor of all three forms of the kinase AKT (AKT1, AKT2, and AKT3) with IC50 values of 58, 210, and 2,200 nM, respectively (Lindsley et al.; Calleja et al.) It displays good selectivity against a panel of 70 other kinases with micromolar inhibition against some kinases, such as calcium/calmodulin-dependent protein kinase 1 and smooth muscle myosin light-chain kinase (Logie et al.)
- Sensitizes prostate tumor and cervical carcinoma cells to apoptotic stimuli (DeFeo-Jones et al.)
- Blocks mitosis and inhibits migration of HeLa cells (Jo et al.)
- Reduces insulin-dependent gene repression in liver cells leading to reduced insulin sensitivity (Logie et al.)
Specifications
Specifications
| CAS | 612847-09-3 |
| Synonym | AKTi-1/2; AKT 1/2 inhibitor |
| Quantity | 10 mg |
| Molecular Formula | C34H29N7O |
| Purity | ≥98% |