missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AKAP9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11386313
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386313 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11386313 Supplier Invitrogen™ Supplier No. PA518345

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

The A-kinase anchor proteins are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternate splicing of this gene results in at least two isoforms that localize to the centrosome and the Golgi apparatus, and interact with numerous signaling proteins from multiple signal transduction pathways. These signaling proteins include type II protein kinase A, serine/threonine kinase protein kinase N, protein phosphatase 1, protein phosphatase 2a, protein kinase C-epsilon and phosphodiesterase 4D3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP9
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene AKAP9
Gene Accession No. Q99996
Gene Alias 5730481H23Rik; A kinase (PRKA) anchor protein (yotiao) 9; A kinase (PRKA) anchor protein 9; AKAP 120-like protein; AKAP 350; AKAP 450; AKAP350; AKAP450; AKAP9; AKAP-9; AKAP9-BRAF fusion protein; A-kinase anchor protein 350 kDa; A-kinase anchor protein 450 kDa; A-kinase anchor protein 9; A-kinase anchoring protein 9; AW545847; C79026; centrosome- and Golgi-localized PKN-associated protein; centrosome- and golgi-localized protein kinase N-associated protein; CG-NAP; G1-448-15; HYPERION; KIAA0803; kinase N-associated protein; LQT11; mei2-5; mKIAA0803; MU-RMS-40.16A; PPP1R45; PRKA9; Protein hyperion; protein kinase A anchoring protein 9; protein kinase A-anchoring protein 9; protein phosphatase 1, regulatory subunit 45; protein yotiao; repro12; YOTIAO
Gene Symbols AKAP9
Host Species Goat
Immunogen Synthetic peptide sequence (SGSTTQFHAGMR) corresponding to the C-terminus amino acids of AKAP9 (aa 3895-3907).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10142
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.