missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1411
This item is not returnable.
View return policy
Description
AKAP3 Polyclonal specifically detects AKAP3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| AKAP3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| A kinase (PRKA) anchor protein 3, AKAP110AKAP 110, AKAP-3, A-kinase anchor protein 110 kDa, Cancer/testis antigen 82, CT82A-kinase anchor protein 3, Fibrous sheath protein of 95 kDa, fibrousheathin 1, Fibrousheathin I, Fibrousheathin-1, FSP95A-kinase anchor protein, 110kDa, PRKA3, protein kinase A anchoring protein 3, Protein kinase A-anchoring protein 3, SOB1fibrous sheath protein, 95kDa, Sperm oocyte-binding protein, sperm oocyte-binding protein 1 | |
| Goat | |
| 95 kDa | |
| 0.1 mg | |
| Primary | |
| Expected from sequence similarity: Mouse, Rat, Dog, Pig, Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| O75969 | |
| AKAP3 | |
| Peptide with sequence C-TPLQLLDWLMVNL corresponding to C-Terminus according to NP_006413.2. | |
| Affinity Purified | |
| RUO | |
| 10566 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction