Learn More
Thermo Scientific™ Air Filter Replacement Kits for Ultra Low Temperature Freezers
Product Code. p-4530860 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
HERAfreeze™ HFU-T, Revco™ UxF, Thermo Scientific™ TSU Series
Revco™ CxF, HERAfreeze™ HFC, TSC-Series 12.7, 17.0 and 20.0 cu. ft. Chest ULT Freezers
Revco™, TS-Series 3.0 cu. ft. Chest ULT Freezers
TDE Series Chest Freezers
TSX Series, Revco RLE, Forma 89000, TLE Series, HERAfreeze HLE
Unit Size:
Each
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11903947 Revco™, TS-Series 3.0 cu. ft. Chest ULT Freezers Pack of 5
11800563 Revco™ CxF, HERAfreeze™ HFC, TSC-Series 12.7, 17.0 and 20.0 cu. ft. Chest ULT Freezers Pack of 5
11346381 HERAfreeze™ HFU-T, Revco™ UxF, Thermo Scientific™ TSU Series Each
15708557 TSX Series, Revco RLE, Forma 89000, TLE Series, HERAfreeze HLE Each
17828010 TDE Series Chest Freezers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11903947 Supplier Thermo Scientific™ Supplier No. 195947
Please to purchase this item. Need a web account? Register with us today!
Easily replace the air filters on your ultra-low temperature freezer with these replacement parts.
Easily replace the air filters on your ultra-low temperature freezer with Thermo Scientific™ Air Filter Replacement Kits for Ultra Low Temperature Freezers.
Replacement air filter
TRUSTED_SUSTAINABILITY
Specifications
Description For use with Forma, HERAfreeze, Revco and TS-Series 3.0 cu. ft. Chest ULT Freezers
Type Freezer Air Filter
Capacity (English) 3 cu. ft.
Capacity (Metric) 84.9 L
Style Chest
For Use With (Equipment) Revco™, TS-Series 3.0 cu. ft. Chest ULT Freezers