Learn More
Air Cadet Nitrile Diaphragm Service Kit for All Pumps
Product Code. 11896155 Shop All Cole-Parmer Products
Change view
Click to view available options
For Use With:
For All Air Cadet Pumps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11896155 For All Air Cadet Pumps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11896155 Supplier Air Cadet Supplier No. 0753006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Nitrile Diaphragm Service Kit for use with all Air Cadet Pumps.
  • Dual-head pump gives you increased capacity over the single-head AIR CADET pumps.
  • Use the two pump heads independently or connect them in series with 3/8 in. ID Tygonr vacuum tubing for up to 23 in. Hg vacuum.
  • Dual-head pump includes thread tape made of PTFE PTFE resin, four polyethylene 3/8 in. NPT(M) x 3/8 in. hose barb adapters, and a 6-ft, three-wire cord.
  • 115 VAC models include a plug.
Specifications
For Use With For All Air Cadet Pumps
Type Nitrile Diaphragm Service Kit