Description
- Dual-head pump gives you increased capacity over the single-head AIR CADET pumps.
- Use the two pump heads independently or connect them in series with 3/8 in. ID Tygonr vacuum tubing for up to 23 in. Hg vacuum.
- Dual-head pump includes thread tape made of PTFE PTFE resin, four polyethylene 3/8 in. NPT(M) x 3/8 in. hose barb adapters, and a 6-ft, three-wire cord.
- 115 VAC models include a plug.
Specifications
Specifications
| For Use With | For All Air Cadet Pumps |
| Type | Nitrile Diaphragm Service Kit |