missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AIDA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555734
This item is not returnable.
View return policy
Description
Immunogen sequence: EEENLEFEED EEEGGAGAGS PDSFPARVPG TLLPRLPSEP GMTLLTIRIE KIGLKDAGQC IDPYITVS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
AIDA acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
Specifications
| AIDA | |
| Polyclonal | |
| Unconjugated | |
| AIDA | |
| 2610208M17Rik; AIDA; Axin interaction partner and dorsalization antagonist; axin interactor, dorsalization associated; axin interactor, dorsalization-associated protein; BC004835; C1orf80 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64853 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96BJ3 | |
| AIDA | |
| Recombinant protein corresponding to Human AIDA. Recombinant protein control fragment (Product #RP-94510). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction