missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AID Monoclonal Antibody (K.15.2)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514921
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
This gene encodes a RNA-editing deaminase that is a member of the cytidine deaminase family. The protein is involved in somatic hypermutation, gene conversion, and class-switch recombination of immunoglobulin genes. Defects in this gene are the cause of autosomal recessive hyper-IgM immunodeficiency syndrome type 2 (HIGM2).
Specifications
| AID | |
| Monoclonal | |
| 40 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q9GZX7 | |
| Aicda | |
| Synthetic peptide corresponding to residues surrounding Leu165 of human AID. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| K.15.2 | |
| Unconjugated | |
| Aicda | |
| activation induced deaminase; activation-induced cytidine deaminase; Aicda; AID; Arp2; CDA2; Cytidine aminohydrolase; epididymis secretory protein Li 284; HEL-S-284; HIGM2; integrated into Burkitt's lymphoma cell line Ramos; single-stranded DNA cytosine deaminase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57379 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction