missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AHSP Polyclonal Antibody
GREENER_CHOICE

Product Code. 17750354
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17750354 200 μL 200µL
17793654 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17750354 Supplier Invitrogen™ Supplier No. PA5118942

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AHSP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen AHSP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene AHSP
Gene Accession No. Q9CY02
Gene Alias AHSP; alpha hemoglobin stabilising protein; alpha hemoglobin stabilizing protein; alpha-hemoglobin-stabilizing protein; Edrf; Eraf; erythroid associated factor; erythroid differentiation associated factor; erythroid differentiation-related factor; Erythroid-associated factor
Gene Symbols AHSP
Host Species Rabbit
Immunogen Recombinant protein Alpha-Hemoglobin Stabilizing Protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170812, 293522
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.