missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ AgilePulse PLUS, Wave Form Generator Only
Click to view available options
Pulse Form:
Square Wave
Unit Size:
Each
Description
Features and Benefits:
- Program up to 5 groups of pulses with separately defined pulse number, voltage, pulse length, interval, and polarity—within each group.
- Direct scale-up from standard low-volume cuvettes to large-volume chambers— transfect up to a billion cells in suspension per sample.
- Incorporate bipolar switching into your electroporation protocols for convenient, more efficient in vivo transfection protocols—pulse chains in each group may be defined as +, -, or flip (+ —> - or - —> +) polarity.
- Maximum number of pulses per protocol increases to 100 pulses.
- Compatible with all BTX specialty electrodes for adherent cell electroporation, intradermal electroporation, intramuscular electroporation and other in vivo transfection applications.
Applications:
- Suspension and Adherent Cell Transfection
- Large Volume Electroporation
- CAR-T Cell Production
- CRISPR Gene Editing
- Intradermal, Intramuscular and Intratumor Delivery
- Animal Immunization
- Vaccine Research
Specifications
Specifications
| For Use With (Application) | Cell Transfection, Electroporation, CAR-T Cell Production, CRISPR Gene Editing |
| For Use With (Equipment) | BTX Specialty Electrodes |
| Product Line | AgilePulse |
| Product Type | Electroporator |
| Voltage | 110/240 V |
| Display | Touchscreen |
| Certifications/Compliance | CE, ETL (UL, CSA), FCC, WEEE, EU RoHS |
| Includes | Waveform Generator Only |
| Pulse Number Range | 1 to 10 Per Group |
| Operating Temperature (Metric) | 4°C to 40°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction