Learn More
STEMCELL Technologies AggreWell™400 24-well Plate Starter Kit
Product Code. 16635655
Encompass_Preferred
Change view
Click to view available options
No. of Wells:
24
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells unitSize
16635655 24 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16635655 Supplier STEMCELL Technologies Supplier No. 34450
Please to purchase this item. Need a web account? Register with us today!
Kit includes 2 AggreWell™400 24-well plates and a bottle of AggreWell™ Rinsing Solution
  • AggreWell™ plates bring an easy, standardized approach to the generation of cell aggregates, including embryoid bodies (EBs) and spheroids
  • EBs and spheroids generated using AggreWell™ plates are consistent in size and shape, and are uniform within and between experiments
  • New and improved second-generation AggreWell™ plates are compatible with a variety of cell types, including ES and iPS cells, cancer cells and more
  • Enhanced optical characteristics provide crystal clear imaging
  • Note: AggreWell™ Rinsing Solution is required for optimal EB and spheroid formation
Specifications
No. of Wells 24
For Use With (Application) Differentiation; Spheroid Culture; Toxicity Assay
Quantity Starter Kit