Learn More
Kinematica Dispersing Aggregate Dia. 20mm PT-DA 20/2MEC-E192
Brand:  Kinematica 11030263
Dispersing Aggregate Dia. 20mm PT-DA 20/2MEC-E192
Product Code. 11990439
  • ONLINE EXCLUSIVE PRICE
    2063.25 €
    1914.00 € / Each
    Save 149.25 €
    7% Off
Only null left
Estimated Shipment: 04-02-2026