missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGAP4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54699
This item is not returnable.
View return policy
Description
AGAP4 Polyclonal specifically detects AGAP4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AGAP4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| AGAP-4, AGAP8, AGAP-8, ARF GTPase-Activating Protein, ArfGAP With GTPase Domain, Ankyrin Repeat And PH Domain 4, ArfGAP With GTPase Domain, Ankyrin Repeat And PH Domain 8, Arf-GAP With GTPase, ANK Repeat And PH Domain-Containing Protein 4, Arf-GAP With GTPase, ANK Repeat And PH Domain-Containing Protein 8, Centaurin, Gamma-Like Family, Member 1, Centaurin, Gamma-Like Family, Member 5, Centaurin-Gamma-Like Family Member 1, CTGLF1, CTGLF5, MRIP2 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 119016 | |
| Human | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| AGAP4 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:PSVSLEFDQQQGSVCPSESEIYEAGA | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction