missing translation for 'onlineSavingsMsg'
Learn More

AG1024, MedChemExpress

Product Code. 16493738
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16493738

Brand: Medchem Express HY10253

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AG1024 (Tyrphostin AG 1024) is a reversible, competitive and selective IGF-1R inhibitor with an IC50 of 7 μM. AG1024 inhibits phosphorylation of IR (IC50=57 μM). AG1024 induces apoptosis and has anti-cancer activity.

AG1024 (Tyrphostin AG 1024) is a reversible, competitive and selective IGF-1R inhibitor with an IC50 of 7 μM. AG1024 inhibits phosphorylation of IR (IC50=57 μM). AG1024 induces apoptosis and has anti-cancer activity.

Specifications

Chemical Name or Material AG1024
CAS 65678-07-1
Color White
Purity Grade Notes Research
Molecular Formula C14H13BrN2O
Quantity 10 mg
Synonym Tyrphostin AG 1024
Solubility Information DMSO : ≥ 50 mg/mL (163.84 mM)
SMILES N#C/C(C#N)=C\C1=CC(C(C)(C)C)=C(O)C(Br)=C1
Molecular Weight (g/mol) 305.17
Formula Weight 305.17
Percent Purity 98.86%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.