Learn More
Aflatoxin P1, TRC
Specifications
Specifications
| Chemical Name or Material | Aflatoxin P1 |
| CAS | 32215-02-4 |
| Molecular Formula | C16 H10 O6 |
| Quantity | 0.5 mg |
| InChI Formula | InChI=1S/C16H10O6/c17-8-2-1-6-11-9(18)5-10-13(7-3-4-20-16(7)21-10)14(11)22-15(19)12(6)8/h3-5,7,16,18H,1-2H2/t7-,16+/m0/s1 |
| SMILES | Oc1cc2O[C@H]3OC=C[C@H]3c2c4OC(=O)C5=C(CCC5=O)c14 |
| IUPAC Name | (3S,7R)-11-hydroxy-6,8,19-trioxapentacyclo[10.7.0.02,9.03,7.013,17]nonadeca-1(12),2(9),4,10,13(17)-pentaene-16,18-dione |
| Molecular Weight (g/mol) | 298.25 |
| Formula Weight | 298.0477 |