Learn More
MedChemExpress Adrenorphin
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Purity | 95.15% |
| CAS | 88377-68-8 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | Cancer-programmed cell death |
| Molecular Weight (g/mol) | 984.18 g/mol |
| Product Type | Reference compound |
| Quantity | 1 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 50 mg/mL (50.80 mM∣Need ultrasonic) |
| Synonym | Metorphamide |