Learn More
MedChemExpress Adrenocorticotropic Hormone (ACTH) (1-39), human
Product Code. 16492038
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492038 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16492038 Supplier MedChemExpress Supplier No. HYP1211
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Adrenocorticotropic Hormone (ACTH) (1-39), human is a melanocortin receptor agonist.
Specifications
Purity 98.07%
CAS 12279-41-3
Content And Storage Protect from light, stored under nitrogen∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
For Use With (Application) Metabolism-protein/nucleotide metabolism
Molecular Weight (g/mol) 4541.14 g/mol
Product Type Peptides
Quantity 5 mg
Shipping Condition Room temperature
Solubility H2O : ≥ 50 mg/mL (11.01 mM)
Synonym 1-39-Corticotropin (human)