Learn More
MedChemExpress Adrenocorticotropic Hormone (ACTH) (1-10), human
Specifications
Specifications
| Purity | 98.53% |
| CAS | 2791-05-1 |
| Content And Storage | Protect from light, stored under nitrogen∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen) |
| For Use With (Application) | Metabolism-protein/nucleotide metabolism |
| Molecular Weight (g/mol) | 1299.41 g/mol |
| Product Type | Peptides |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 250 mg/mL (192.40 mM∣Need ultrasonic) ∣H2O : 1 mg/mL (0.77 mM∣Need ultrasonic) |