missing translation for 'onlineSavingsMsg'
Learn More

Adrenocorticotrophic Hormone (ACTH) Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Adrenocorticotrophic Hormone (ACTH) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-452-P1ABX

 View more versions of this product

Product Code. 12681467

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Host Species: Mouse

Clone: AH26

Isotype: IgG1

Species Reactivity: Human. It shows a broad species reactivity.

Epitope: aa 1-24

Immunogen: A synthetic peptide corresponding to aa 1-24 from the N-terminal of human ACTH

Positive Control: Normal pituitary gland or pituitary adenoma

Cellular Localization: Cytoplasmic Ab-1 is excellent for staining of routine formalin-fixed, paraffin-embedded tissues. It labels corticotrophs in the adenohypophysis and is useful in the classification of pituitary adenomas. NeoMarkers' Ab-1 and Ab-2 may provide a pair for ELISA

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adrenocorticotrophic Hormone (ACTH) Ab-1
Monoclonal
1mg/mL
Mouse
100 μL
Steroids Hormones and Growth Factors
Human
IgG1
Immunohistochemistry (Paraffin)
AH26
Unconjugated
A synthetic peptide corresponding to aa 1-24 from the N-terminal of human ACTH
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.