Learn More
GeneBLAzer™ ADRA2A-Gqo5-NFAT-bla CHO-K1 Cells
GeneBLAzer™ ADRA2A-Gqo5-NFAT-bla CHO-K1 Cells contain the human Adrenoceptor A2A.
Brand:  GeneBLAzer™ K1743
Additional Details : Weight : 0.01000kg
Product Code. 10701624
  • 15770.00 € / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
  • Gene Symbol: ADRA2A
  • Accession Number: NM_000681.2
  • Population Status: Single Cell Clone Origin
  • GeneBLAzer™ ADRA2A-Gqo5-NFAT-bla CHO-K1 cells contain the human Adrenoceptor A2A (ADRA2A) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cells (Cat. no. K1536) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Chimeric G protein, Gqo5.
  • The GeneBLAzer™ ADRA2A-Gqo5-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of UK 14,304. In addition, GeneBLAzer™ ADRA2A-Gqo5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
Specifications
Specifications
Liquid Nitrogen
GPCRs
2 x 106 Cells
Fluorescent
ADRA2A-Gqo5-NFAT-bla CHO-K1 cells, ADRA2A-Gqo5-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis
Dividing Cells