Learn More
GeneBLAzer™ ADRA2A-Gqo5-NFAT-bla CHO-K1 Cells
GeneBLAzer™ ADRA2A-Gqo5-NFAT-bla CHO-K1 Cells contain the human Adrenoceptor A2A.
Brand: GeneBLAzer™ K1743
Additional Details : Weight : 0.01000kg
Description
- Gene Symbol: ADRA2A
- Accession Number: NM_000681.2
- Population Status: Single Cell Clone Origin
- GeneBLAzer™ ADRA2A-Gqo5-NFAT-bla CHO-K1 cells contain the human Adrenoceptor A2A (ADRA2A) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cells (Cat. no. K1536) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Chimeric G protein, Gqo5.
- The GeneBLAzer™ ADRA2A-Gqo5-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of UK 14,304. In addition, GeneBLAzer™ ADRA2A-Gqo5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
Specifications
Liquid Nitrogen | |
GPCRs | |
2 x 106 Cells |
Fluorescent | |
ADRA2A-Gqo5-NFAT-bla CHO-K1 cells, ADRA2A-Gqo5-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
Dividing Cells |