missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADP-Sugar Pyrophosphatase/NUDT5 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99496C
This item is not returnable.
View return policy
Description
ADP-Sugar Pyrophosphatase/NUDT5 Polyclonal specifically detects ADP-Sugar Pyrophosphatase/NUDT5 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ADP-Sugar Pyrophosphatase/NUDT5 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| EC 3.6.1.-, EC 3.6.1.13, hYSAH1, nucleoside diphosphate linked moiety X-type motif 5, Nucleoside diphosphate-linked moiety X motif 5, nudix (nucleoside diphosphate linked moiety X)-type motif 5, Nudix motif 5, YSA1, YSA1HADP-sugar pyrophosphatase | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ADP-Sugar Pyrophosphatase/NUDT5. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 11164 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction