missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADORA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102048
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ADORA3.
Adenosine is involved in a variety of processes, including the synthesis of urea, the anti-inflammatory response and the inhibition of protein synthesis The adenosine receptors, including adenosine A1-R, adenosine A2A-R, adenosine A2B-R and adenosine A3-R, are integral membrane proteins that are members of the G protein-coupled receptor family. The A1-R protein mediates ureagenesis in a partially calcium-dependent manner. Adenosine is known to mediate coronary vasodilation via the A2A-R receptor. Collagen synthesis and total protein synthesis are inhibited in certain cells by adenosine, acting via the A2B receptors. Activation of the A3-R receptor inhibits the induction of the cytokine TNF alpha and blocks the endotoxin CD14 receptor signal transduction pathway.
Specifications
| ADORA3 | |
| Polyclonal | |
| Unconjugated | |
| ADORA3 | |
| A3 adenosine receptor; A3AR; A3R; AA3R; AD026; adenosine A3 receptor; Adenosine A3-R; adenosine receptor A3; adenosine-3 receptor; Adora3; Agtrl1; angiotensin II receptor-like 1; angiotensin receptor-like 1; Apelin receptor; Apj; APJ (apelin) receptor; APJ receptor; APJR; Aplnr; ARA3; B78; BA552M11.5; G protein-coupled receptor; G protein-coupled receptor APJ; Gpcr2; GPCR34; G-protein coupled receptor 2; G-protein coupled receptor APJ; G-protein coupled receptor HG11; HG11; HG11 orphan receptor; MSR; msr/apj; RP11-552M11.7; TGPCR1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11542, 140 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P0DMS8, Q61618 | |
| ADORA3 | |
| A synthesized peptide derived from human ADORA3(Accession P0DMS8), corresponding to amino acid residues Y284-E318. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction