missing translation for 'onlineSavingsMsg'
Learn More

AdipoRon, MedChemExpress

Product Code. 16442937
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16442937

Brand: Medchem Express HY15848

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AdipoRon is an orally active adiponectin receptor (AdipoR) agonist, binding to AdipoR1 and AdipoR2 with Kds of 1.8 and 3.1 μM, respectively.

AdipoRon is an orally active adiponectin receptor (AdipoR) agonist, binding to AdipoR1 and AdipoR2 with Kds of 1.8 and 3.1 μM, respectively.

Specifications

Chemical Name or Material AdipoRon
CAS 924416-43-3
Color White
Purity Grade Notes Research
Molecular Formula C27H28N2O3
Quantity 50 mg
Solubility Information DMSO : ≥ 44 mg/mL (102.68 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NC1CCN(CC2=CC=CC=C2)CC1)COC3=CC=C(C(C4=CC=CC=C4)=O)C=C3
Molecular Weight (g/mol) 428.52
Formula Weight 428.52
Percent Purity 96.13%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.