missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adiponectin Recombinant Superclonal™ Antibody
Adiponectin Recombinant Polyclonal Antibody
Brand: Invitrogen 710179
This item is not returnable.
View return policy
Description
Adiponectin Recombinant Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| Adiponectin | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q15848, Q60994 | |
| ADIPOQ | |
| Recombinant protein corresponding to amino acids 57-244 of human adiponectin. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 11HCLC | |
| Unconjugated | |
| ADIPOQ | |
| 30 kDa adipocyte complement-related protein; 30kDa; Acdc; Acrp30; Ad; adipo; adipocyte complement related protein; adipocyte complement related protein of 30 kDa; adipocyte complement-related 30 kDa protein; adipocyte complement-related protein of 30 kDa; adipocyte, C1Q and collagen domain containing; adipocyte, C1q and collagen domain-containing protein; Adipocyte-specific protein AdipoQ; adiponectin; adiponectin, C1Q and collagen domain containing; Adipoq; Adipose most abundant gene transcript 1 protein; adipose specific collagen-like factor; ADIPQTL1; ADPIOQ; ADPN; Apm1; APM-1; APN; GBP28; Gelatin-binding protein; gelatin-binding protein 28 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11450, 246253, 9370 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction