Learn More
Invitrogen™ Adiponectin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1054
Description
PA1-054 detects Adiponectin (30 kDa) from mouse serum. PA1-054 has been successfully used in Western blot and immunoprecipitation procedures. By Western blot, this antibody detects an ~30 kDa protein representing Adiponectin from mouse serum. The PA1-054 immunogen is a pair of synthetic peptides corresponding to the residues E(18) D D V T T T E E L A P A L V(32) and F(187) T Y D Q Y Q E K N V D Q A(200) of mouse Adiponectin.
Adiponectin is a protein hormone secreted exclusively by adipocytes that regulates the metabolism of lipids and glucose. Adiponectin has been shown to play a role in various physiological processes including homeostasis and obesity. Decreased levels of adiponectin are associated with obese individuals as well as insulin resistance and hyperinsulinemia (type 2 diabetes). In mice adiponectin has been shown to improve insulin signaling, thus ameliorating insulin resistance. Through activation of AMP kinase and PPAR-alpha, adiponectin can increase fatty acid transport, oxidation, and dissipation in skeletal muscle, leading to a reduction of intramyocellular lipid levels and thus improved insulin signaling.
Specifications
| Adiponectin | |
| Polyclonal | |
| Unconjugated | |
| ADIPOQ | |
| 30 kDa adipocyte complement-related protein; 30kDa; Acdc; Acrp30; Ad; adipo; adipocyte complement related protein; adipocyte complement related protein of 30 kDa; adipocyte complement-related 30 kDa protein; adipocyte complement-related protein of 30 kDa; adipocyte, C1Q and collagen domain containing; adipocyte, C1q and collagen domain-containing protein; Adipocyte-specific protein AdipoQ; adiponectin; adiponectin, C1Q and collagen domain containing; Adipoq; Adipose most abundant gene transcript 1 protein; adipose specific collagen-like factor; ADIPQTL1; ADPIOQ; ADPN; Apm1; APM-1; APN; GBP28; Gelatin-binding protein; gelatin-binding protein 28 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11450 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| Q60994 | |
| ADIPOQ | |
| Synthetic peptides corresponding to the residues E(18) D D V T T T E E L A P A L V(32) and F(187) T Y D Q Y Q E K N V D Q A(200) of mouse Adiponectin. | |
| 200 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.