Learn More
ADH4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16474-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes class II alcohol dehydrogenase 4 pi subunit, which is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Class II alcohol dehydrogenase is a homodimer composed of 2 pi subunits. It exhibits a high activity for oxidation of long-chain aliphatic alcohols and aromatic alcohols and is less sensitive to pyrazole. This gene is localized to chromosome 4 in the cluster of alcohol dehydrogenase genes.Specifications
| ADH4 | |
| Polyclonal | |
| Unconjugated | |
| ADH4 | |
| ADH 2, ADH4, Alcohol dehydrogenase 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 127, 26876, 29646 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08319, Q64563, Q9QYY9 | |
| ADH4 | |
| ADH4 Fusion Protein Ag9568 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |