missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Adenylate Cyclase 5/6 Polyclonal Antibody

Product Code. 16637764
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16637764 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16637764 Supplier Invitrogen™ Supplier No. PA5101284

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ADCY5/6.

Adenylate cyclase 5/6 encodes a member of the membrane-bound adenylyl cyclase enzymes. Adenylyl cyclases mediate G protein-coupled receptor signaling through the synthesis of the second messenger cAMP. Activity of the encoded protein is stimulated by the Gs alpha subunit of G protein-coupled receptors and is inhibited by protein kinase A, calcium and Gi alpha subunits. Single nucleotide polymorphisms in this gene may be associated with low birth weight and type 2 diabetes. Alternatively spliced transcript variants that encode different isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase 5/6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADCY6
Gene Accession No. O43306, O95622, P84309, Q01341, Q03343, Q04400
Gene Alias A cyclase V; A cyclase VI; AC5; AC6; ACVI; Adcy5; Adcy6; ADCYB; adenylate cyclase 5; adenylate cyclase 6; adenylate cyclase type 5; Adenylate cyclase type 6; adenylate cyclase type V; Adenylate cyclase type VI; adenylyl cyclase 5; Adenylyl cyclase 6; adenylyl cyclase type V; ATP pyrophosphate-lyase 5; ATP pyrophosphate-lyase 6; AW121902; ca(2+)-inhibitable adenylyl cyclase; FDFM; KIAA0422; LCCS8; mKIAA0422
Gene Symbols ADCY5, ADCY6
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADCY5(Accession O95622), corresponding to amino acid residues Q867-S917.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 111, 112, 11512, 224129, 25289, 64532
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.