Learn More
Adenosine deaminase, MedChemExpress
Description
Adenosine deaminase is an enzyme that catalyzes the irreversible deamination of adenosine and 2'-deoxyadenosine to inosine and 2'-deoxyinosine, respectively.
Specifications
Specifications
| Chemical Name or Material | Adenosine deaminase |
| Physical Form | Powder |
| CAS | 9026-93-1 |
| Color | White |
| Quantity | 1 mg |
| Purity Grade Notes | Research |
| For Use With (Application) | COVID-19-immunoregulation |
| SMILES | [Adenosine deaminase] |
| Grade | Research |