Learn More
Adenosine A2aR Antibody (Ig2838) - Chimeric - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-09003-0.2mg
 View more versions of this product
Product Code. 18604037
  • 591.00 € / 0.20mg
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Adenosine A2aR Monoclonal antibody specifically detects Adenosine A2aR in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adenosine A2aR
Monoclonal
1 mg/mL
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
ELISA, Immunofluorescence, Neutralization
Ig2838
Unconjugated
adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8
Human recombinant A2-adenosine receptor.
0.2 mg
Apoptosis, GPCR, Immunology, Innate Immunity
135
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ