Learn More
Adavivint, MedChemExpress
Product Code. 16433016
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16433016
Brand: Medchem Express HY109049
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Adavivint (SM04690; Lorecivivint) is a potent and selective inhibitor of canonical Wnt signaling, with an EC50 of 19.5 nM via a high-throughput TCF/LEF-reporter assay in SW480 colon cancer cells.
Adavivint (SM04690
Specifications
Chemical Name or Material Adavivint
CAS 1467093-03-3
Color Gray
Purity Grade Notes Research
Molecular Formula C29H24FN7O
Quantity 25 mg
Synonym SM04690 Lorecivivint
Solubility Information DMSO : 25 mg/mL (49.45 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC(C)CC(NC1=CC(C2=CC3=C(NN=C3C(N4)=NC5=C4C(C6=CC=CC(F)=C6)=CN=C5)C=C2)=CN=C1)=O
Molecular Weight (g/mol) 505.55
Formula Weight 505.55
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation