missing translation for 'onlineSavingsMsg'
Learn More

Adavivint, MedChemExpress

Product Code. 16430597
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16430597

Brand: Medchem Express HY109049

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Adavivint (SM04690; Lorecivivint) is a potent and selective inhibitor of canonical Wnt signaling, with an EC50 of 19.5 nM via a high-throughput TCF/LEF-reporter assay in SW480 colon cancer cells.

Adavivint (SM04690

Specifications

Chemical Name or Material Adavivint
CAS 1467093-03-3
Color Gray
Purity Grade Notes Research
Molecular Formula C29H24FN7O
Quantity 50 mg
Synonym SM04690 Lorecivivint
Solubility Information DMSO : 25 mg/mL (49.45 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC(C)CC(NC1=CC(C2=CC3=C(NN=C3C(N4)=NC5=C4C(C6=CC=CC(F)=C6)=CN=C5)C=C2)=CN=C1)=O
Molecular Weight (g/mol) 505.55
Formula Weight 505.55
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.