Learn More
Thermo Scientific™ Adapters for Oval Single Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Choice of 4 adapters to accommodate a range of blood bags in oval single blood bag buckets for Thermo Scientific™ HAEMAFlex™ 6/8 rotors.
402.00 € - 482.00 €
Specifications
For Use With (Equipment) Oval single blood bag buckets
View More Specs
Products 3
Product Code Brand Description Capacity Price Quantity & Availability  
Product Code Brand Description Capacity Price Quantity & Availability  
16679312
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
75003838
400-450mL Double or Triple Blood Bag Adapter (set of 2) 6/8 x 450 mL
472.00 €
Pack of 2
View alternative products
missing translation for 'toSaveUpto' 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16574551
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
75003839
250mL Platelet-rich Plasma or Buffy Coat Bag Adapter (set of 2) 6/8 x 250 mL
402.00 €
Each
View alternative products
missing translation for 'toSaveUpto' 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16564551
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
75003837
450-550mL Quad or Quint Blood Bag Adapter (set of 2) 6/8 x 550 mL
482.00 €
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 09-09-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Thermo Scientific™ blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.
Select adapters for 450–550mL Quad or Quint blood bags, 400–450mL Double or Triple blood bags, 250mL Platelet-rich Plasma or Buffy Coat bags or 400–450mL Single or Double blood bags.
Specifications
Specifications
Oval single blood bag buckets