Learn More
Ibidi Adapter for Sticky-Slide I Luer
Click to view available options
Quantity:
1
Unit Size:
Each
Description
- For secure fastening of sticky-Slides to coverslips in combination with the Clamp for sticky-Slides (80040)
Specifications
Specifications
| Type | Sticky-slide Adapter |
| Quantity | 1 |