Learn More
ADAPTER M1/4'-28F X 1,6MM ID ETFE
Brand:  Kimble Chase 953918-2306
Product Code. 11812923
  • ONLINE EXCLUSIVE PRICE
    25.45 €
    12.05 € / Each
    Save 13.40 €
    52% Off
Only null left
Estimated Shipment: 03-03-2026