Learn More
Eppendorf™ Adapter For LC 481 TwinTec PCR Plate
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
For LC 481 TwinTec PCR Plate
Unit Size:
Each
Description
- One-piece design achieved using twin-shot technology, an innovative injection moulding process
- Minimal bending of skirts
- Semi-skirted for labeling and bar-coding
- Suitable for quantitative real-time PCR
- Fits most Thermal Cyclers
- 250μL max. well volume (when used with cap strips)
Specifications
Specifications
| Type | Adapter |
| For Use With (Equipment) | For LC 481 TwinTec PCR Plate |
| Quantity | 1 Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion